J.T.Baker® BAKERBOND Speedisk® C18, SPE Columns
Supplier: Avantor
7606-02EA 511.42 EUR
7606-02 7606-06 7606-09 7606-11
J.T.Baker® BAKERBOND Speedisk® C18, SPE Columns
Chromatography Columns
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Non ionic, non polar to moderately polar analytes.
These SPE columns can be used in reversed phase mode.