J.T.Baker® BAKERBOND Speedisk® Silica, SPE Columns
Supplier: Avantor
8163-02EA 0 EUR
8163-02 8163-06 8163-11
J.T.Baker® BAKERBOND Speedisk® Silica, SPE Columns
Chromatography Columns
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Adsorbs polar analytes from non polar solvents like hydrocarbons and less polar esters and ethers.
These SPE columns can be used in adsorbtion mode.