1-Bromo-3,3-dimethylbutanone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone
MQ200
414131-25ML 414131-5ML
SIAL414131-25MLEA 171 EUR
SIAL414131-25ML SIAL414131-5ML
1-Bromo-3,3-dimethylbutanone, Sigma-Aldrich®
1-Bromo-3,3-dimethylbutanone
Formula: C₆H₁₁BrO
MW: 179.06 g/mol
Boiling Pt: 47…49 °C (1 mmHg)
Melting Pt: 76
Density: 1.3508 g/cm³ (0 °C)
Storage Temperature: Freezer
MDL Number: MFCD00000206
CAS Number: 5469-26-1
EINECS: 226-794-0
UN: 2810
ADR: 6.1 II