2,2-Bis(bromomethyl)-1,3-propanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dibromoneopentyl glycol, 2,2-Bis(bromomethyl)propane-1,3-diol
M-Clarity™ quality level = MQ100
301930-1KG 301930-25G 301930-3KG
SIAL301930-1KGEA 114 EUR
SIAL301930-1KG SIAL301930-25G SIAL301930-3KG
2,2-Bis(bromomethyl)-1,3-propanediol, Sigma-Aldrich®
2,2-Bis(bromomethyl)-1,3-propanediol
Formula: C₅H₁₀Br₂O₂
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 107…109 °C
Density: 2.2 g/cm³ (20 °C)
MDL Number: MFCD00004688
CAS Number: 3296-90-0
EINECS: 221-967-7