2-Ethylbutyraldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
110094-1L 110094-250ML
SIAL110094-1LEA 397 EUR
SIAL110094-1L SIAL110094-250ML
2-Ethylbutyraldehyde, Sigma-Aldrich®
2-Ethylbutyraldehyde
Formula: C₆H₁₂O
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.8306 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006985
CAS Number: 97-96-1
EINECS: 202-623-5
UN: 1178
ADR: 3,II