Isobutyraldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Methylpropanal, 2-Methylpropionaldehyde
M-Clarity™ quality level = MQ200
240788-100ML 240788-2ML
SIAL240788-100MLEA 74.2 EUR
SIAL240788-100ML SIAL240788-2ML
Isobutyraldehyde, Sigma-Aldrich®
Isobutyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 64
Melting Pt: –65 °C
Density: 0.794 g/cm³ (20 °C)
Flash Pt: –24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006980
CAS Number: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3 II
Merck Index: 13,05171