1-Chloro-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
178004-100ML 178004-500ML
SIAL178004-100MLEA 102 EUR
SIAL178004-100ML SIAL178004-500ML
1-Chloro-2-methylpropane, Sigma-Aldrich®
1-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68
Melting Pt: –131 °C
Density: 0.8829 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000954
CAS Number: 513-36-0
EINECS: 208-157-9
UN: 1127
ADR: 3 II
Merck Index: 13,05155