2-Chloro-2-methylbutane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

277029-100G 277029-25G
SIAL277029-100GEA 243 EUR
SIAL277029-100G SIAL277029-25G
2-Chloro-2-methylbutane, Sigma-Aldrich®
2-Chloro-2-methylbutane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –73 °C
Density: 0.8591 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000818
CAS Number: 594-36-5
EINECS: 209-836-2
UN: 1107
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR