2-Methyl-3-pentanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethyl Isopropyl ketone
M-Clarity™ quality level = MQ200
108707-25G 108707-5G
SIAL108707-25GEA 189 EUR
SIAL108707-25G SIAL108707-5G
2-Methyl-3-pentanone, Sigma-Aldrich®
2-Methyl-3-pentanone
Formula: C₆H₁₂O
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –69 °C
Density: 0.8845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009314
CAS Number: 565-69-5
EINECS: 209-288-4
UN: 1224
ADR: 3,II