3-Methyl-2-pentanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
M67001-25G M67001-5G
SIALM67001-25GEA 213 EUR
SIALM67001-25G SIALM67001-5G
3-Methyl-2-pentanone, Sigma-Aldrich®
3-Methyl-2-pentanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 117 °C (1013 hPa)
Density: 0.815 g/cm³ (20 °C)
MDL Number: MFCD00009336
CAS Number: 565-61-7
EINECS: 209-282-1
UN: 1224
ADR: 3,II