4-Hydroxy-4-methyl-2-pentanone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Diacetone alcohol
M-Clarity™ quality level = MQ200
H41544-2.5L H41544-25ML H41544-500ML H41544-4L
SIALH41544-2.5LEA 66.6 EUR
SIALH41544-2.5L SIALH41544-25ML SIALH41544-500ML SIALH41544-4L
4-Hydroxy-4-methyl-2-pentanone, Sigma-Aldrich®
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM