2,2,6,6-Tetramethyl-3,5-heptanedione, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dipivaloylmethane, TMHD, 2,2,6,6-Tetramethylheptane-3,5-dione
M-Clarity™ quality level = MQ200
155756-100G 155756-25G 155756-5G
SIAL155756-100GEA 249 EUR
SIAL155756-100G SIAL155756-25G SIAL155756-5G
2,2,6,6-Tetramethyl-3,5-heptanedione, Sigma-Aldrich®
2,2,6,6-Tetramethyl-3,5-heptanedione
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3