Pivalic anhydride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Trimethylacetic anhydride, 2,2-Dimethylpropionic anhydride
M-Clarity™ quality level = MQ200
143502-100G
143502-25G
SIAL143502-100GEA
134
EUR
SIAL143502-100G
SIAL143502-25G
Pivalic anhydride, Sigma-Aldrich®
Pivalic anhydride
Formula:
C₁₀H₁₈O₃ MW: 186.25 g/mol Boiling Pt: 193 °C (1013 hPa) Density: 0.91 g/cm³ (20 °C) Flash Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008842 CAS Number: 1538-75-6 EINECS: 216-263-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...