Pivalamide
Supplier: Thermo Scientific
Synonyms:
Trimethylacetamide, 2,2,2-Trimethylacetamide
TL00202.1
TL00202.50
MAYBTL00202.1EA
0
EUR
MAYBTL00202.1
MAYBTL00202.50
Pivalamide
Pivalamide
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 212 °C (1013 hPa) Density: 1,02 g/cm³ (20 °C) |
MDL Number:
MFCD00008011 CAS Number: 754-10-9 EINECS: 212-043-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...