L(-)-Glutathione (reduced form), Sigma-Aldrich®
Supplier: Merck
Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
M-Clarity™ quality level = MQ300
G4251-5G G4251-1G G4251-10MG G4251-100G G4251-10G G4251-250G G4251-25G G4251-300MG G4251-500G G4251-50G
SIALG4251-5GEA 62.9 EUR
SIALG4251-5G SIALG4251-1G SIALG4251-10MG SIALG4251-100G SIALG4251-10G SIALG4251-250G SIALG4251-25G SIALG4251-300MG SIALG4251-500G SIALG4251-50G
L(-)-Glutathione (reduced form), Sigma-Aldrich®
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488