L(-)-Glutathione (reduced form), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
G6013-25G G6013-100G G6013-10G G6013-5G
SIALG6013-25GEA 535 EUR
SIALG6013-25G SIALG6013-100G SIALG6013-10G SIALG6013-5G
L(-)-Glutathione (reduced form), Sigma-Aldrich®
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488