L(-)-Glutathione (oxidised form), Sigma-Aldrich®
Supplier: Merck
Synonyms: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
MQ300
G4376-10G G4376-1G G4376-250MG G4376-500MG G4376-5G
SIALG4376-10GEA 473 EUR
SIALG4376-10G SIALG4376-1G SIALG4376-250MG SIALG4376-500MG SIALG4376-5G
L(-)-Glutathione (oxidised form), Sigma-Aldrich®
L(-)-Glutathione (oxidized form)
Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612.64 g/mol
Melting Pt: 185
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8
EINECS: 248-170-7