L(-)-Glutathione (oxidised form), Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALG4501-100MGEA 43.1 EUR
SIALG4501-100MG SIALG4501-1G SIALG4501-250MG SIALG4501-500MG SIALG4501-5G
L(-)-Glutathione (oxidised form), Sigma-Aldrich®
L(-)-Glutathione (oxidised form)
Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612,64 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00063106
CAS Number: 27025-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR