2,2-Dimethylbutyric acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,2-Dimethylbutanoic acid
M-Clarity™ quality level = MQ100
D152609-100ML D152609-25ML
SIALD152609-100MLEA 128 EUR
SIALD152609-100ML SIALD152609-25ML
2,2-Dimethylbutyric acid, Sigma-Aldrich®
2,2-Dimethylbutyric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 74 °C (0.5 mmHg)
Melting Pt: –13 °C
Density: 0.9449 g/cm³ (0 °C)
MDL Number: MFCD00004200
CAS Number: 595-37-9
EINECS: 209-865-0
UN: 3265
ADR: 8,III