Isobutyric acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Methylpropionic acid, 2-Methylpropanoic acid
M-Clarity™ quality level = MQ100
I1754-100ML I1754-1L I1754-500ML
SIALI1754-100MLEA 36.6 EUR
SIALI1754-100ML SIALI1754-1L SIALI1754-500ML
Isobutyric acid, Sigma-Aldrich®
Isobutyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 13,05172