Isovaleric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid
M-Clarity™ quality level = MQ300
W310212-100G-K W310212-1KG-K W310212-5KG-K
SIALW310212-100G-KEA 76.9 EUR
SIALW310212-100G-K SIALW310212-1KG-K SIALW310212-5KG-K
Isovaleric acid, Sigma-Aldrich®
Isovaleric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 177 °C (1013 hPa)
Melting Pt: –33 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002726
CAS Number: 503-74-2
EINECS: 207-975-3
UN: 3265
ADR: 8,II
Merck Index: 12,05251