4-Methylvaleric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isocaproic acid, 4-Methylpentanoic acid
M-Clarity™ quality level = MQ100
277827-25G 277827-5G
SIAL277827-25GEA 76 EUR
SIAL277827-25G SIAL277827-5G
4-Methylvaleric acid, Sigma-Aldrich®
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200
Melting Pt: –35 °C
Density: 0.9192 g/cm³ (20 °C)
Flash Pt: 89
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0