4-Methylvaleric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isocaproic acid, 4-Methylpentanoic acid
M-Clarity™ quality level = MQ300
W346306-10KG-K W346306-1KG-K W346306-250G-K
SIALW346306-10KG-KEA 3060 EUR
SIALW346306-10KG-K SIALW346306-1KG-K SIALW346306-250G-K
4-Methylvaleric acid, Sigma-Aldrich®
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.9192 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0