3,3-Dimethylglutaric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid
D4379-100G D4379-250G D4379-25G
SIALD4379-100GEA 154 EUR
SIALD4379-100G SIALD4379-250G SIALD4379-25G
3,3-Dimethylglutaric acid, Sigma-Aldrich®
3,3-Dimethylglutaric acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 98…101 °C
Density: 1.4278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7
EINECS: 225-425-0