2-Iodo-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
Synonyms: tert-Butyl iodide, 2-Methyl-2-propyliodide
M-Clarity™ quality level = MQ100
245798-100G 245798-25G
SIAL245798-100GEA 387 EUR
SIAL245798-100G SIAL245798-25G
2-Iodo-2-methylpropane, Sigma-Aldrich®
2-Iodo-2-methylpropane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –78 °C
Density: 1.55 g/cm³ (20 °C)
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001068
CAS Number: 558-17-8
EINECS: 209-190-1
UN: 2391
ADR: 3,II