2-Ethylhexylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Ethyl-1-hexylamine
M-Clarity™ quality level = MQ200
E29508-100ML E29508-1L
SIALE29508-100MLEA 27.8 EUR
SIALE29508-100ML SIALE29508-1L
2-Ethylhexylamine, Sigma-Aldrich®
2-Ethylhexylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 169.2
Melting Pt: –76 °C
Density: 0.7882 g/cm³ (20 °C)
Flash Pt: 50
Storage Temperature: Ambient
MDL Number: MFCD00008148
CAS Number: 104-75-6
EINECS: 203-233-8
UN: 2276
ADR: 3 III