Isobutylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Amino-2-methylpropane, 2-Methylpropylamine
MQ200
I14150-100ML I14150-1L I14150-500ML I14150-5ML
SIALI14150-100MLEA 33.1 EUR
SIALI14150-100ML SIALI14150-1L SIALI14150-500ML SIALI14150-5ML
Isobutylamine, Sigma-Aldrich®
Isobutylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 66
Melting Pt: –85 °C
Density: 0.7344 g/cm³ (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008146
CAS Number: 78-81-9
EINECS: 201-145-4
UN: 1214
ADR: 3 II
Merck Index: 13,05149