Isopentylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Amino-3-methylbutane, Isopentyl amine, Isoamylamine, 3-Methylbutylamine, Leucamine
M-Clarity™ quality level = MQ100
126810-100G 126810-10G
SIAL126810-100GEA 171 EUR
SIAL126810-100G SIAL126810-10G
Isopentylamine, Sigma-Aldrich®
Isopentylamine
Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.75 g/cm³ (20 °C)
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008203
CAS Number: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3,II
Merck Index: 13,05128