tert-Butylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Aminoisobutane
M-Clarity™ quality level = MQ200
391433-100ML 391433-1L
SIAL391433-100MLEA 86.8 EUR
SIAL391433-100ML SIAL391433-1L
tert-Butylamine, Sigma-Aldrich®
tert-Butylamine
Formula: C₄H₁₁N
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.6965 g/cm³ (20 °C)
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544