tert-Butylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Aminoisobutane
M-Clarity™ quality level = MQ300
B89205-100ML B89205-1L B89205-2.5L B89205-500ML B89205-5ML
SIALB89205-100MLEA 26 EUR
SIALB89205-100ML SIALB89205-1L SIALB89205-2.5L SIALB89205-500ML SIALB89205-5ML
tert-Butylamine, Sigma-Aldrich®
tert-Butylamine
Formula: C₄H₁₁N
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.6965 g/cm³ (20 °C)
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544