AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP
M-Clarity™ quality level = MQ200
08581-1L
08581-250ML
SIAL08581-1LEA
67.5
EUR
SIAL08581-1L
SIAL08581-250ML
AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
AMP (2-Amino-2-methylpropanol)
Formula:
(CH₃)₂C(NH₂)CH₂OH MW: 89.14 g/mol Boiling Pt: 165 Melting Pt: 18…26 °C Density: 0.667 g/cm³ (20 °C) Flash Pt: 67 Storage Temperature: Ambient |
MDL Number:
MFCD00008051 CAS Number: 124-68-5 EINECS: 204-709-8 Merck Index: 13,00448 |