AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP
M-Clarity™ quality level = MQ200
A9199-100ML A9199-250ML A9199-500ML
SIALA9199-100MLEA 77.8 EUR
SIALA9199-100ML SIALA9199-250ML SIALA9199-500ML
AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
AMP (2-Amino-2-methylpropanol)
Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165
Melting Pt: 18…26 °C
Density: 0.667 g/cm³ (20 °C)
Flash Pt: 67
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448