Diisobutylamine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
135186-1L
135186-250ML
SIAL135186-1LEA
71.1
EUR
SIAL135186-1L
SIAL135186-250ML
Diisobutylamine, Sigma-Aldrich®
Diisobutylamine
Formula:
C₈H₁₉N Boiling Pt: 137 °C (1013 hPa) Melting Pt: –73.5 °C Density: 0.763 g/cm³ (0 °C) |
MDL Number:
MFCD00008930 CAS Number: 110-96-3 EINECS: 203-819-3 UN: 2361 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...