2,4-Dimethyl-3-pentanone ≥98%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,4-Dimethylpentan-3-one, Diisopropyl ketone, Isobutyrone
M-Clarity™ quality level = MQ100
136867-100ML 136867-500ML
SIAL136867-100MLEA 47.9 EUR
SIAL136867-100ML SIAL136867-500ML
2,4-Dimethyl-3-pentanone ≥98%, Sigma-Aldrich®
2,4-Dimethyl-3-pentanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.8445 g/cm³ (0 °C)
MDL Number: MFCD00008918
CAS Number: 565-80-0
EINECS: 209-993-7
UN: 1987
ADR: 3,III