2-Ethyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
E14659-100ML E14659-500ML
SIALE14659-100MLEA 61.3 EUR
SIALE14659-100ML SIALE14659-500ML
2-Ethyl-1-butanol, Sigma-Aldrich®
2-Ethyl-1-butanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: < –15 °C
Density: 0.8233 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004744
CAS Number: 97-95-0
EINECS: 202-621-4
UN: 1987
ADR: 3,III