Isobutanol

Supplier: EHRENSTORFER GMBH

EHERC15142000EA 45.9 EUR
EHERC15142000
Isobutanol
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74,12 g/mol
Boiling Pt: 107,9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0,802 g/cm³ (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00004740
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://si.vwr.com/stibo/web/std.lang.all/12/85/2301285.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR