2-Methyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol
M-Clarity™ quality level = MQ200
152463-1L
152463-2.5L
152463-250ML
SIAL152463-1LEA
111
EUR
SIAL152463-1L
SIAL152463-2.5L
SIAL152463-250ML
2-Methyl-2-butanol, Sigma-Aldrich®
2-Methyl-2-butanol
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 102 °C (1013 hPa) Melting Pt: –8.4 °C Density: 0.806 g/cm³ (20 °C) Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004478 CAS Number: 75-85-4 EINECS: 200-908-9 UN: 1105 ADR: 3,II Merck Index: 13,07218 |