3,3-Dimethyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,3-Dimethylbutan-1-ol
M-Clarity™ quality level = MQ100
183105-10G 183105-50G
SIAL183105-10GEA 127 EUR
SIAL183105-10G SIAL183105-50G
3,3-Dimethyl-1-butanol, Sigma-Aldrich®
3,3-Dimethyl-1-butanol
Formula: C₆H₁₄O
Boiling Pt: 142…143 °C (1013 hPa)
Melting Pt: –65…–60 °C
Density: 0.844 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002928
CAS Number: 624-95-3
EINECS: 210-872-6
UN: 1987
ADR: 3,III