Isoamyl alcohol, Sigma-Aldrich®

Supplier: Merck

SIAL59085-250MLEA 64.7 EUR
SIAL59085-250ML
Isoamyl alcohol, Sigma-Aldrich®
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88,15 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0,812 g/cm³ (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00002934
CAS Number: 123-51-3
ADR: 3,III
Merck Index: 13,05214
https://si.vwr.com/stibo/web/std.lang.all/15/69/2301569.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR