2,2-Dimethyl-1,3-propanediol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol
M-Clarity™ quality level = MQ100
538256-25G 538256-3KG 538256-500G
SIAL538256-25GEA 28 EUR
SIAL538256-25G SIAL538256-3KG SIAL538256-500G
2,2-Dimethyl-1,3-propanediol, Sigma-Aldrich®
2,2-Dimethyl-1,3-propanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 208
Melting Pt: 128…130 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004685
CAS Number: 126-30-7
EINECS: 204-781-0
Merck Index: 12,06547