2,3-Dimethylpentane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D173207-100G D173207-25G D173207-5G
SIALD173207-100GEA 558 EUR
SIALD173207-100G SIALD173207-25G SIALD173207-5G
2,3-Dimethylpentane, Sigma-Aldrich®
2,3-Dimethylpentane
Formula: C₇H₁₆
MW: 100.2 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –135 °C
Density: 0.6956 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009337
CAS Number: 565-59-3
EINECS: 209-280-0
UN: 1206
ADR: 3,II