2,4-Dimethylpentane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
D173401-25G D173401-5G
SIALD173401-25GEA 246 EUR
SIALD173401-25G SIALD173401-5G
2,4-Dimethylpentane, Sigma-Aldrich®
2,4-Dimethylpentane
Formula: C₇H₁₆
MW: 100.2 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: –119.2 °C
Density: 0.6734 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008945
CAS Number: 108-08-7
EINECS: 203-548-0
UN: 1206
ADR: 3,II