Squalane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosane
M-Clarity™ quality level = MQ200
234311-100G 234311-500G 234311-5G
SIAL234311-100GEA 59.7 EUR
SIAL234311-100G SIAL234311-500G SIAL234311-5G
Squalane, Sigma-Aldrich®
Squalane
Formula: C₃₀H₆₂
MW: 422.82 g/mol
Boiling Pt: 470
Melting Pt: –35 °C
Density: 0.758 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3
EINECS: 203-825-6
Merck Index: 12,08923