3,3-Dimethylpentane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

118451-1G 118451-5G
SIAL118451-1GEA 66.7 EUR
SIAL118451-1G SIAL118451-5G
3,3-Dimethylpentane, Sigma-Aldrich®
3,3-Dimethylpentane
Formula: C₇H₁₆
MW: 100.2 g/mol
Boiling Pt: 86 °C (1013 hPa)
Melting Pt: –135 °C
Density: 0.6936 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009322
CAS Number: 562-49-2
EINECS: 209-230-8
UN: 1206
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR