3-Methylpentane, Sigma-Aldrich®
Supplier: Merck
Danger
MQ100
M66005-100ML M66005-500ML M66005-5ML
SIALM66005-100MLEA 58.4 EUR
SIALM66005-100ML SIALM66005-500ML SIALM66005-5ML
3-Methylpentane, Sigma-Aldrich®
3-Methylpentane
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 64
Melting Pt: –118 °C
Density: 0.664 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009342
CAS Number: 96-14-0
EINECS: 202-481-4
UN: 1208
ADR: 3 II