(α,α,α-Trifluoro-2-tolyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-(Trifluoromethyl)phenylacetic acid, (2-(Trifluoromethyl)phenyl)acetic acid
M-Clarity™ quality level = MQ100
235296-1G
235296-5G
SIAL235296-1GEA
28.5
EUR
SIAL235296-1G
SIAL235296-5G
(α,α,α-Trifluoro-2-tolyl)acetic acid, Sigma-Aldrich®
(α,α,α-Trifluoro-2-tolyl)acetic acid
Formula:
F₃CC₆H₄CH₂COOH MW: 204.15 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 98…100 °C |
MDL Number:
MFCD00004327 CAS Number: 3038-48-0 EINECS: 221-240-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...