2-Naphthylacetic acid ≥99%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid
A13528.06 A13528.09 A13528.18
A13528.06EA 50.3 EUR
A13528.06 A13528.09 A13528.18
2-Naphthylacetic acid ≥99%
2-Naphthylacetic acid
Purity: 99% CAS No.: 581-96-4 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 141-143° Beilstein Reference: 973396
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 141…143 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0