2-Naphthylacetic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

MQ100

N4002-25G-A N4002-5G-A
SIALN4002-25G-AEA 101 EUR
SIALN4002-25G-A SIALN4002-5G-A
2-Naphthylacetic acid, Sigma-Aldrich®
2-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352
Melting Pt: 139…142 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR