(2-Bromophenyl)acetic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-(2-Bromophenyl)acetic acid
296650100 296650050
ACRO296650100EA 30.2 EUR
ACRO296650100 ACRO296650050
(2-Bromophenyl)acetic acid 98%
(2-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 105 to 110 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %