(4-Chlorophenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid
M-Clarity™ quality level = MQ100
139262-100G 139262-10G
SIAL139262-100GEA 81 EUR
SIAL139262-100G SIAL139262-10G
(4-Chlorophenyl)acetic acid, Sigma-Aldrich®
(4-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 101…104 °C
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6